Få mere at vide
(S)-4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol, TRC
Artikelnummer. 30004888
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30004888 2.5 mg 2.5mg
30012528 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004888 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3237902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 197712-01-9
Molekylær formel C10 H15 N3 O2
Molekylvægt (g/mol) 209.245
Synonym (αS)-α-[3-(Methylnitrosoamino)propyl3-pyridinemethanol, (S)-α-[3-(Methylnitrosoamino)propyl]-3-pyridinemethanol, (S)-NNAL
IUPAC navn N-[(4S)-4-hydroxy-4-(3-pyridyl)butyl]-N-methyl-nitrous amide
SMIL CN(CCC[C@H](O)c1cccnc1)N=O
Tekniske data
Kemisk navn eller materiale (S)-4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol
Mængde 2.5 mg
InChI formel InChI=1S/C10H15N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8,10,14H,3,5,7H2,1H3/t10-/m0/s1
Formel vægt 209.1164