Få mere at vide
(S)-4-[(Methoxycarbonyl)amino]-5-hexenoic Acid Ethyl Ester, TRC
Artikelnummer. 30473726
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473726 5 mg
30478728 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473726 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2263685MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-4-[(Methoxycarbonyl)amino]-5-hexenoic Acid Ethyl Ester
CAS 148073-34-1
Molekylær formel C10H17NO4
Mængde 5 mg
Synonym (S)-Ethyl 4-((methoxycarbonyl)amino)hex-5-enoate
InChI formel InChI=1S/C10H17NO4/c1-4-8(11-10(13)14-3)6-7-9(12)15-5-2/h4,8H,1,5-7H2,2-3H3,(H,11,13)/t8-/m1/s1
SMIL CCOC(=O)CC[C@H](NC(=O)OC)C=C
IUPAC navn ethyl (4S)-4-(methoxycarbonylamino)hex-5-enoate
Molekylvægt (g/mol) 215.25
Formel vægt 215.1158