missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Boc-(3-hydroxymethyl)morpholine, 97%, Thermo Scientific™

Catalog Number p-7055878
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
714971-28-5
C10H19NO4
217.265
MFCD06799478
AIQSXVGBMCJQAG-QMMMGPOBSA-N
s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate, s-4-boc-3-hydroxymethyl morpholine, s-n-boc-3-hydroxymethyl morpholine, tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate, s-n-boc-3-hydroxymethylmorpholine, 3 s-hydroxymethyl-4-bocmorpholine, s-4-boc-3-hydroxymethylmorpholine, s-4-boc-3-hydroxymethyl-morpholine, s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem5634
1514266
tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
CC(C)(C)OC(=O)N1CCOCC1CO
This item is not returnable. View return policy
714971-28-5
C10H19NO4
217.265
MFCD06799478
AIQSXVGBMCJQAG-QMMMGPOBSA-N
s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate, s-4-boc-3-hydroxymethyl morpholine, s-n-boc-3-hydroxymethyl morpholine, tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate, s-n-boc-3-hydroxymethylmorpholine, 3 s-hydroxymethyl-4-bocmorpholine, s-4-boc-3-hydroxymethylmorpholine, s-4-boc-3-hydroxymethyl-morpholine, s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem5634
1514266
tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
CC(C)(C)OC(=O)N1CCOCC1CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
An intermediate of BMS-599626, a selective and orally efficacious inhibitor of human epidermal growth factor receptor 1 and 2 kinases.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 80°C (literature)
Color White
Beilstein 10251196
Solubility Information Slightly soluble in water.
Formula Weight 217.27
Percent Purity 97%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material (S)-4-Boc-(3-hydroxymethyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'