Learn More
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%, Thermo Scientific Chemicals
Catalog Number 15408486
Click to view available options
Quantity:
250 mg
1 g
5 g
252049-08-4
C27H28N2O5
460.53
MFCD02259487
KKUPBFMKUPMFRE-VWLOTQADSA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
70700347
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid
C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
252049-08-4
C27H28N2O5
460.53
MFCD02259487
KKUPBFMKUPMFRE-VWLOTQADSA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
70700347
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid
C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 474.51
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid