Få mere at vide
(S)-4-Benzyl-3-propionyl-2-oxazolidinone-d3, TRC
Artikelnummer. 30341525
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30341525 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341525 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28865250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-4-Benzyl-3-propionyl-2-oxazolidinone-d3
CAS 156451-08-0
Molekylær formel C13H12D3NO3
Mængde 50 mg
Synonym (4S)-3-(1-Oxopropyl)-4-(phenylmethyl)-2-oxazolidinone-d3, (+)-4-Benzyl-3-propionyl-2-oxazolidinone-d3, (4S)-4-Benzyl-3-propanoyloxazolidin-2-one-d3, (4S)-4-Benzyl-3-propionyl-1,3-oxazolidin-2-one-d3, (S)-N-Propionyl-4-benzyl-1,3-oxazolidin-2-one-d3, 3-[1-Oxopropyl]-4(S)-(phenylmethyl)-2-oxazolidinone-d3
InChI formel InChI=1S/C13H15NO3/c1-2-12(15)14-11(9-17-13(14)16)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3/t11-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])CC(=O)N1[C@@H](Cc2ccccc2)COC1=O
IUPAC navn (4S)-4-benzyl-3-(3,3,3-trideuteriopropanoyl)-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 236.28
Formel vægt 236.124