Learn More
(S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 78287-27-1 |
| Molecular Formula | C22H20N2O4 |
| MDL Number | MFCD06657922 |
| Quantity | 250 mg |
| SMILES | CCC1=C2CN3C(=CC4=C(COC(=O)[C@]4(O)CC)C3=O)C2=NC2=CC=CC=C12 |
| Molecular Weight (g/mol) | 376.41 |
| Percent Purity | 98% |
| Chemical Name or Material | (S)-4,11-Diethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione |