Få mere at vide
(S)-3-Methylheptanoic Acid-d3, TRC
Artikelnummer. 30466855
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30466855 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466855 Leverandør Toronto Research Chemicals Leverandørnr. TRCM311937500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-3-Methylheptanoic Acid-d3
CAS 1329647-33-7
Molekylær formel C8H13D3O2
Mængde 500 mg
Synonym (S)-(-)-3-Methylheptanoic Acid-d3, (S)-3-Methyl-heptanoic Acid-d3
InChI formel InChI=1S/C8H16O2/c1-3-4-5-7(2)6-8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/t7-/m0/s1/i2D3
SMIL [2H]C([2H])([2H])[C@@H](CCCC)CC(=O)O
IUPAC navn (3S)-3-(trideuteriomethyl)heptanoic acid
Molekylvægt (g/mol) 147.23
Formel vægt 147.1339