missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-3-Hydroxybutanoic acid, MedChemExpress

Artikelnummer. 16495457
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16495457 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16495457 Leverandør Medchem Express Leverandørnr. HYW050031

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress (S)-3-Hydroxybutanoic acid is a normal human metabolite, that has been found elevated in geriatric patients remitting from depression. In humans, 3-Hydroxybutyric acid is synthesized in the liver from acetyl-CoA, and can be used as an energy source by the brain when blood glucose is low.

(S)-3-Hydroxybutanoic acid is a normal human metabolite, that has been found elevated in geriatric patients remitting from depression. In humans, 3-Hydroxybutyric acid is synthesized in the liver from acetyl-CoA, and can be used as an energy source by the brain when blood glucose is low.

Tekniske data

Kemisk navn eller materiale (S)-3-Hydroxybutanoic acid
CAS 6168-83-8
Farve White
Renhedsgrad noter Research
Molekylær formel C4H8O3
Mængde 50 mg
Synonym (S)-β-Hydroxybutanoic acid L-(+)-3-Hydroxybutyric acid L-β-Hydroxybutyric acid
Opløselighedsinformation H2O : 100 mg/mL (960.61 mM; Need ultrasonic)
SMIL C[C@H](O)CC(O)=O
Molekylvægt (g/mol) 104.1
Formel vægt 104.1
Procent renhed 97.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.