Få mere at vide
(S)-3’-Hydroxy-N-methylcoclaurine Hydrochloride (~75% ee), TRC
Artikelnummer. 30133573
Skift visning
Mængde:
0.5 mg
1 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133573 10 mg
30004932 0.5 mg
30008478 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133573 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94649510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 138644-99-2
Molekylær formel C18H22ClNO4
Molekylvægt (g/mol) 351.8
Synonym (S)-4-[(1,2,3,4-Tetrahydro-7-hydroxy-6-methoxy-2-methyl-1-isoquinolinyl)methyl]-1,2-benzenediol Hydrochloride
IUPAC navn (S)-4-((7-hydroxy-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-1-yl)methyl)benzene-1,2-diol hydrochloride
SMIL OC1=CC=C(C[C@@H]2N(C)CCC3=C2C=C(O)C(OC)=C3)C=C1O.Cl
Tekniske data
Kemisk navn eller materiale (S)-3’-Hydroxy-N-methylcoclaurine Hydrochloride
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C18H21NO4.ClH/c1-19-6-5-12-9-18(23-2)17(22)10-13(12)14(19)7-11-3-4-15(20)16(21)8-11;/h3-4,8-10,14,20-22H,5-7H2,1-2H3;1H/t14-;/m0./s1
Formel vægt 351.12
Procent renhed >95%