missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-3-Benzyloxycarbonyl-4-(3-oxobutyl)-5-oxazilidinone, TRC

Product Code. 30335202
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30335202 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335202 Supplier Toronto Research Chemicals Supplier No. TRCB28592050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (S)-3-Benzyloxycarbonyl-4-(3-oxobutyl)-5-oxazilidinone
CAS 89662-51-1
Molekylær formel C15 H17 N O5
Mængde 50 mg
InChI formel InChI=1S/C15H17NO5/c1-11(17)7-8-13-14(18)21-10-16(13)15(19)20-9-12-5-3-2-4-6-12/h2-6,13H,7-10H2,1H3/t13-/m0/s1
SMIL CC(=O)CC[C@@H]1N(COC1=O)C(=O)OCc2ccccc2
IUPAC navn benzyl (4S)-5-oxo-4-(3-oxobutyl)-1,3-oxazolidine-3-carboxylate
Molekylvægt (g/mol) 291.3
Formel vægt 291.1107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.