Få mere at vide
(S)-3-Amino-2-azepanone (>90%), TRC
Artikelnummer. 30317256
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30317256 50 mg
1 options
Artikelnummer. 30317256 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60410350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-3-Amino-2-azepanone
CAS 21568-87-6
Molekylær formel C6H12N2O
Mængde 50 mg
Synonym (3S)-3-Aminohexahydro-2H-azepin-2-one, (-)-3-Aminohexahydro-2-azepinone, (-)-alpha-Amino-epsilon-caprolactam, (-)-alpha-Aminocaprolactam, (3S)-Amino-epsilon-caprolactam, (S)-(-)-alpha-Amino-epsilon-caprolactam, (S)-3-Amino-2-caprolactam, (S)-3-Aminoazepan-2-one, (S)-3-Aminohexahydro-2-azepinone, (S)-Caprolactam-3-amine, (S)-alpha-Amino-epsilon-caprolactam, (S)-alpha-Aminocaprolactam, L-(-)-alpha-Amino-epsilon-caprolactam, L-alpha-Amino-epsilon-caprolactam, L-alpha-Aminocaprolactam
InChI formel InChI=1S/C6H12N2O/c7-5-3-1-2-4-8-6(5)9/h5H,1-4,7H2,(H,8,9)/t5-/m0/s1
SMIL N[C@H]1CCCCNC1=O
IUPAC navn (3S)-3-aminoazepan-2-one
Molekylvægt (g/mol) 128.17