Få mere at vide
(S)-3-Amino-1-(tert-butoxycarbonyl)piperidine-3-carboxylicacid, TRC
Artikelnummer. 30412963
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412963 500 mg
30410773 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30412963 Leverandør Toronto Research Chemicals Leverandørnr. TRCS296850500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-3-Amino-1-(tert-butoxycarbonyl)piperidine-3-carboxylicacid
CAS 862372-92-7
Molekylær formel C11 H20 N2 O4
Mængde 500 mg
InChI formel InChI=1S/C11H20N2O4/c1-10(2,3)17-9(16)13-6-4-5-11(12,7-13)8(14)15/h4-7,12H2,1-3H3,(H,14,15)/t11-/m0/s1
SMIL CC(C)(C)OC(=O)N1CCC[C@](N)(C1)C(=O)O
IUPAC navn (3S)-3-amino-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
Molekylvægt (g/mol) 244.287
Formel vægt 244.142