Få mere at vide
Tocris Bioscience™ S 2101
LSD1 inhibitor
Brand: Tocris Bioscience™ 5714/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
S 2101 is a LSD1 inhibitor (IC50 = 990 nM; Ki = 610 nM). Exhibits selectivity for LSD1 over MAO-B and MAO-A (Ki values are 17 and 110 μM, respectively).Tekniske data
| (1R,2S)-rel-2-[3,5-Difluoro-2-(phenylmethoxy)phenyl]cycloprpanamine hydrochloride | |
| 1239262-36-2 | |
| 50 mg |