Få mere at vide
(S)-2-Methylvaleric Acid, TRC
Artikelnummer. 30479168
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479168 10 mg
30474177 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479168 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33852010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1187-82-2
Molekylær formel C6H12O2
Molekylvægt (g/mol) 116.16
Synonym (+)-2-Methylpentanoic Acid, (+)-2-Methylvaleric Acid, (S)-(+)-2-Methylpentanoic Acid, (S)-(+)-2-Methylvaleric Acid, (S)-2-Methylpentanoic Acid
IUPAC navn (2S)-2-methylpentanoic acid
SMIL CCCC(C)C(=O)O
Tekniske data
Kemisk navn eller materiale (S)-2-Methylvaleric Acid
Mængde 10 mg
InChI formel InChI=1S/C6H12O2/c1-3-4-5(2)6(7)8/h5H,3-4H2,1-2H3,(H,7,8)/t5-/m0/s1
Formel vægt 116.0837