Få mere at vide
(S)-2-Methylhexanol-d3, TRC
Artikelnummer. 30475092
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30475092 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475092 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3119571G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2-Methylhexanol-d3
CAS 1329614-76-7
Molekylær formel C7H13D3O
Mængde 1 g
Synonym (S)-2-Methyl-1-hexanol-d3, (-)-2-Methyl-1-hexanol-d3
InChI formel InChI=1S/C7H16O/c1-3-4-5-7(2)6-8/h7-8H,3-6H2,1-2H3/t7-/m0/s1/i2D3
SMIL [2H]C([2H])([2H])[C@H](CO)CCCC
IUPAC navn (2S)-2-(trideuteriomethyl)hexan-1-ol
Molekylvægt (g/mol) 119.22
Formel vægt 119.1389