missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-(+)-2-Methylbutyric Acid-d3, TRC

Artikelnummer. 30465703
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465703 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465703 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29435625MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (S)-(+)-2-Methylbutyric Acid-d3
CAS 175779-05-2
Molekylær formel C5H7D3O2
Mængde 25 mg
Synonym (2S)-2-Methyl-butanoic Acid-d3, (+)-2-Methylbutanoic Acid-d3, (+)-2-Methylbutyric Acid-d3, (+)-alpha-Methylbutyric Acid-d3, (2S)-2-Methylbutanoic Acid-d3, (2S)-2-Methylbutyric Acid-d3, (S)-2-Methylbutanoic Acid-d3, (S)-2-Methylbutyric Acid-d3, S-(+)-2-Methylbutanoic Acid-d3
InChI formel InChI=1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/t4-/m0/s1/i2D3
SMIL [2H]C([2H])([2H])[C@@H](CC)C(=O)O
IUPAC navn (2S)-2-(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 105.15
Formel vægt 105.0869
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.