missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals

Catalog Number p-3779058
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
112022-81-8, 108-88-3
C18H20BNO
277.17
MFCD00078439
VMKAFJQFKBASMU-KRWDZBQOSA-N
s-2-methyl-cbs-oxazaborolidine, s-methyl oxazaborolidine, s---2-methyl-cbs-oxazaborolidine, s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, corey catalyst, s-methyl-cbs-oxazaborolidine, s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, s-me-cbs catalyst, s-methyl-cbs, s-me-cbs
2734713
(3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
[H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
112022-81-8, 108-88-3
C18H20BNO
277.17
MFCD00078439
VMKAFJQFKBASMU-KRWDZBQOSA-N
s-2-methyl-cbs-oxazaborolidine, s-methyl oxazaborolidine, s---2-methyl-cbs-oxazaborolidine, s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, corey catalyst, s-methyl-cbs-oxazaborolidine, s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, s-me-cbs catalyst, s-methyl-cbs, s-me-cbs
2734713
(3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
[H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 28.34, 67.18
CAS Max % 32.82, 71.66
Density 0.9290g/mL
Packaging Glass bottle
Quantity 5 mL
Formula Weight 277.17
Chemical Name or Material (S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (S)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'