Få mere at vide
(S)-2-Methyl-1-(4-toluenesulfonyloxy)hexane-d3, TRC
Artikelnummer. 30468575
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30468575 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468575 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3302921G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2-Methyl-1-(4-toluenesulfonyloxy)hexane-d3
CAS 1329610-58-3
Molekylær formel C14H19D3O3S
Mængde 1 g
Synonym (2S)-2-(Methyl-d3)-1-hexanol 4-Methylbenzenesulfonate, (S)-2-(Methyl-d3)-1-hexyl Tosylate
InChI formel InChI=1S/C14H22O3S/c1-4-5-6-13(3)11-17-18(15,16)14-9-7-12(2)8-10-14/h7-10,13H,4-6,11H2,1-3H3/t13-/m0/s1/i3D3
SMIL [2H]C([2H])([2H])[C@@H](CCCC)COS(=O)(=O)c1ccc(C)cc1
IUPAC navn [(2S)-2-(trideuteriomethyl)hexyl] 4-methylbenzenesulfonate
Molekylvægt (g/mol) 273.41
Formel vægt 273.1478