missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Diphenylmethylpyrrolidine, 97+%, Thermo Scientific Chemicals

Catalog Number p-3757780
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
119237-64-8
C17H20N
238.35
MFCD00799525
OXOBKZZXZVFOBB-INIZCTEOSA-O
s-2-benzhydrylpyrrolidine, s---2-diphenylmethyl pyrrolidine, s-2-diphenylmethylpyrrolidine, 2s-2-diphenylmethyl pyrrolidine, 2s-2-benzhydrylpyrrolidine, 2s-2-benzylhydrylpyrrolidine, 2r-2-benzhydrylpyrrolidine, pubchem18168, 2alpha-benzhydrylpyrrolidine, s-2-benzhydryl-pyrrolidine
2734053
C1C[NH2+][C@@H](C1)C(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
119237-64-8
C17H20N
238.35
MFCD00799525
OXOBKZZXZVFOBB-INIZCTEOSA-O
s-2-benzhydrylpyrrolidine, s---2-diphenylmethyl pyrrolidine, s-2-diphenylmethylpyrrolidine, 2s-2-diphenylmethyl pyrrolidine, 2s-2-benzhydrylpyrrolidine, 2s-2-benzylhydrylpyrrolidine, 2r-2-benzhydrylpyrrolidine, pubchem18168, 2alpha-benzhydrylpyrrolidine, s-2-benzhydryl-pyrrolidine
2734053
C1C[NH2+][C@@H](C1)C(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0620g/mL
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.5860 to 1.5880
Quantity 250 mg
Specific Gravity 1.062
Specific Rotation Condition - 3.00 (25.00°C C=1,Chloroform)
Specific Rotation − 3.00
IUPAC Name (2S)-2-benzhydrylpyrrolidine
Formula Weight 237.34
Percent Purity 97+%
Physical Form Oil
Chemical Name or Material (S)-2-Diphenylmethylpyrrolidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-2-Diphenylmethylpyrrolidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'