missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol, 99%, Thermo Scientific Chemicals

Catalog Number p-7046309
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
111060-52-7
C23H25NO
331.46
MFCD00191984
ZXNVOFMPUPOZDF-UHFFFAOYNA-N
s-+-2-dibenzylamino-3-phenyl-1-propanol, s-2-dibenzylamino-3-phenylpropan-1-ol, 2s-2-dibenzylamino-3-phenylpropan-1-ol, dibenzyl phenylalaninol, n,n-dibenzyl-l-phenylalaninol, 2-s-dibenzylamino-3-phenylpropan-1-ol, s-beta-dibenzylamino benzene-1-propanol, s--2-dibenzylamino-3-phenyl-1-propanol, benzenepropanol,b-bis phenylmethyl amino-, bs
7019180
(2S)-2-(dibenzylamino)-3-phenylpropan-1-ol
OCC(CC1=CC=CC=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1
This item is not returnable. View return policy
111060-52-7
C23H25NO
331.46
MFCD00191984
ZXNVOFMPUPOZDF-UHFFFAOYNA-N
s-+-2-dibenzylamino-3-phenyl-1-propanol, s-2-dibenzylamino-3-phenylpropan-1-ol, 2s-2-dibenzylamino-3-phenylpropan-1-ol, dibenzyl phenylalaninol, n,n-dibenzyl-l-phenylalaninol, 2-s-dibenzylamino-3-phenylpropan-1-ol, s-beta-dibenzylamino benzene-1-propanol, s--2-dibenzylamino-3-phenyl-1-propanol, benzenepropanol,b-bis phenylmethyl amino-, bs
7019180
(2S)-2-(dibenzylamino)-3-phenylpropan-1-ol
OCC(CC1=CC=CC=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 72°C to 74°C
Quantity 1 g
Beilstein 7253058
Optical Rotation +8.5° (c=4 in Methanol)
Formula Weight 331.46
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'