missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2-Chloropropionic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.
Solubility
Fully miscible in water.
Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.
Solubility
Fully miscible in water.
Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 4°C |
Density | 1.27 |
Boiling Point | 187°C |
Flash Point | 107°C (225°F) |
Refractive Index | 1.4345 |
Quantity | 5 g |
UN Number | UN2511 |
Beilstein | 1720257 |
Solubility Information | Fully miscible in water. |
Formula Weight | 108.53 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'