Få mere at vide
(S)-(-)-2-Chloro-3-[4(5)-imidazolyl]propionic Acid, TRC
Artikelnummer. 30361671
Skift visning
Mængde:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361671 100 mg
30355713 10 mg
30357721 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30361671 Leverandør Toronto Research Chemicals Leverandørnr. TRCC366870100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17561-26-1
Molekylær formel C6 H7 Cl N2 O2
Molekylvægt (g/mol) 174.58
IUPAC navn (2S)-2-chloro-3-(1H-imidazol-4-yl)propanoic acid
SMIL OC(=O)[C@@H](Cl)Cc1c[nH]cn1
Tekniske data
Kemisk navn eller materiale (S)-(-)-2-Chloro-3-[4(5)-imidazolyl]propionic Acid
Mængde 100 mg
InChI formel InChI=1S/C6H7ClN2O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1H2,(H,8,9)(H,10,11)/t5-/m0/s1
Formel vægt 174.0196