missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Bromopropionic acid, 98%, >85% ee, Thermo Scientific Chemicals

Catalog Number p-3777710
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
32644-15-8
C3H5BrO2
152.98
MFCD00137325
MONMFXREYOKQTI-REOHCLBHSA-N
s---2-bromopropionic acid, s-2-bromopropionic acid, 2s-2-bromopropanoic acid, s-2-bromopropanoic acid, l-2-bromopropionic acid, s---2-bromopropionicacid, l-alpha-bromopropionic acid, 2s---2-bromopropionic acid, s-alpha-bromopropionic acid
642232
(2S)-2-bromopropanoic acid
CC(C(=O)O)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
32644-15-8
C3H5BrO2
152.98
MFCD00137325
MONMFXREYOKQTI-REOHCLBHSA-N
s---2-bromopropionic acid, s-2-bromopropionic acid, 2s-2-bromopropanoic acid, s-2-bromopropanoic acid, l-2-bromopropionic acid, s---2-bromopropionicacid, l-alpha-bromopropionic acid, 2s---2-bromopropionic acid, s-alpha-bromopropionic acid
642232
(2S)-2-bromopropanoic acid
CC(C(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -35°C
Color Colorless
Density 1.7000g/mL
Boiling Point 203°C
Flash Point 100°C
Enantiomeric Excess 80% min. (Typically)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CH(Br)CO2H
Refractive Index 1.4740 to 1.476
Quantity 5 g
Beilstein 02, 254
Specific Gravity 1.7
Specific Rotation -27
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 152.98
Percent Purity 98%,>85% ee
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Bromopropionic acid, >85%ee
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'