Få mere at vide
(S)-2-Amino-5-methoxytetralin (S)-Mandelate, Mikromol™
Artikelnummer. 30462576
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462576 4 x 25 mg
30050805 100 mg
30049653 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462576 Leverandør Mikromol Leverandørnr. MM1403.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(S)-2-Amino-5-methoxytetralin (S)-Mandelate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2-Amino-5-methoxytetralin (S)-Mandelate
CAS 439133-67-2
Analyt- eller komponentnavne (S)-2-Amino-5-methoxytetralin (S)-Mandelate
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N O . C8 H8 O3
Certificeringer/compliance ISO 17025
Synonym (S)-2-Amino-5-methoxytetralin (S)-Mandelate, (aS)-(2S)-compd. with 1,2,3,4-Tetrahydro-5-methoxy-2-naphthalenamine a-Hydroxybenzeneacetic acid, compd. with (2S)-1,2,3,4-Tetrahydro-5-methoxy-2-naphthalenamine (aS)-a-Hydroxybenzeneacetic acid
InChI formel InChI=1S/C11H15NO.C8H8O3/c1-13-11-4-2-3-8-7-9(12)5-6-10(8)11;9-7(8(10)11)6-4-2-1-3-5-6/h2-4,9H,5-7,12H2,1H3;1-5,7,9H,(H,10,11)/t9-;7-/m00/s1
Løsningstype Neat
SMIL COc1cccc2C[C@@H](N)CCc12.O[C@H](C(=O)O)c3ccccc3
IUPAC navn (2S)-2-hydroxy-2-phenylacetic acid;(2S)-5-methoxy-1,2,3,4-tetrahydronaphthalen-2-amine
Molekylvægt (g/mol) 329.39
Mængde 4 x 25 mg
Formel vægt 329.1627 g/mol