Få mere at vide
S-(+)-2-Amino-1-propanol-3,3,3-d3, TRC
Artikelnummer. 30315738
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315738 1 mg
30327716 10 mg
30323164 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315738 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6281271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S)-2-Aminopropan-1-ol-3,3,3-D3
CAS 352438-84-7
Molekylær formel C3 D3 H6 N O
Mængde 1 mg
Synonym 1-Propan-3,3,3-d3-ol, 2-amino-, (2S)- (9CI), (2S)-2-Aminopropan-1-ol-3,3,3-D3, L-Alaninol-d3, (2S)-2-Amino-1-propan-3,3,3-d3-ol
InChI formel InChI=1S/C3H9NO/c1-3(4)2-5/h3,5H,2,4H2,1H3/t3-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])[C@H](N)CO
IUPAC navn (2S)-2-amino-3,3,3-trideuteriopropan-1-ol
Molekylvægt (g/mol) 78.1282
Formel vægt 78.0872