Få mere at vide
(S)-(+)-2-Amino-1-propanol-3,3,3-d3, CDN
Artikelnummer. 30314014
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314014 0.05 g
30311422 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314014 Leverandør CDN Leverandørnr. CDND51640.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2 S)-2-Aminopropan-1-ol-3,3,3-D3
CAS 352438-84-7
Molekylær formel C3 D3 H6 N O
Mængde 0.05 g
Synonym 1-Propan-3,3,3-d3-ol, 2-amino-, (2 S)- (9 CI), (2 S)-2-Aminopropan-1-ol-3,3,3-D3, L-Alaninol-d3, (2 S)-2-Amino-1-propan-3,3,3-d3-ol
InChI formel InChI=1 S/C3H9NO/c1-3(4)2-5/h3,5 H,2,4H2,1H3/t3-/m0/s1/i1D3
SMIL [2 H]C([2 H])([2 H])[C@H](N)CO
IUPAC navn (2 S)-2-amino-3,3,3-trideuteriopropan-1-ol
Molekylvægt (g/mol) 78.1282
Formel vægt 78.0872 g/mol
Procent renhed 99 atom % D, min 97% Chemical Purity