Få mere at vide
(S)-2-Acetamido-5-ureidopentanoic Acid, TRC
Artikelnummer. 30135021
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135021 100 mg
30011791 50 mg
30010983 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135021 Leverandør Toronto Research Chemicals Leverandørnr. TRCA790148100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 33965-42-3
Molekylær formel C8H15N3O4
Molekylvægt (g/mol) 217.22
Synonym Nalpha-Acetyl-L-citrulline, alpha-N-Acetylcitrulline, N2-Acetyl-N5-(aminocarbonyl)-L-ornithine, (2S)-2-Acetamido-5-(carbamoylamino)pentanoic Acid
IUPAC navn (2S)-2-acetamido-5-(carbamoylamino)pentanoic acid
SMIL CC(=O)N[C@@H](CCCNC(=O)N)C(=O)O
Tekniske data
Kemisk navn eller materiale (S)-2-Acetamido-5-ureidopentanoic Acid
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H15N3O4/c1-5(12)11-6(7(13)14)3-2-4-10-8(9)15/h6H,2-4H2,1H3,(H,11,12)(H,13,14)(H3,9,10,15)/t6-/m0/s1
Mængde 100 mg
Formel vægt 217.1063
Procent renhed >95%