missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propionic acid, TRC

Artikelnummer. 30393140
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30393140 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30393140 Leverandør Toronto Research Chemicals Leverandørnr. TRCF58869050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Safinamide Acid
CAS 1160513-60-9
Molekylær formel C17 H18 F N O3
Mængde 50 mg
Synonym L-Alanine, N-[[4-[(3-fluorophenyl)methoxy]phenyl]methyl]-, N-[[4-[(3-Fluorophenyl)methoxy]phenyl]methyl]-L-alanine, (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propionic acid, Safinamide free acid, Safinamide Acid
InChI formel InChI=1S/C17H18FNO3/c1-12(17(20)21)19-10-13-5-7-16(8-6-13)22-11-14-3-2-4-15(18)9-14/h2-9,12,19H,10-11H2,1H3,(H,20,21)/t12-/m0/s1
SMIL C[C@H](NCc1ccc(OCc2cccc(F)c2)cc1)C(=O)O
IUPAC navn (2S)-2-[[4-[(3-fluorophenyl)methoxy]phenyl]methylamino]propanoic acid
Molekylvægt (g/mol) 303.328
Formel vægt 303.127
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.