Få mere at vide
(S)-2,2-Dimethyl-1,3-dioxolane-4-methanol-d2, TRC
Artikelnummer. 30379924
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30379924 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379924 Leverandør Toronto Research Chemicals Leverandørnr. TRCD472636100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S)-2,2-Dimethyl-1,3-dioxolane-4-methanol-d2
CAS 110220-54-7
Molekylær formel C6H10D2O3
Mængde 100 mg
Synonym (S)-Glycerol-3,3-d2-1,2-acetonide, (4S)-2,2-Dimethyl-1,3-dioxolane-4-methan-alpha,alpha-d2-ol
InChI formel InChI=1S/C6H12O3/c1-6(2)8-4-5(3-7)9-6/h5,7H,3-4H2,1-2H3/t5-/m0/s1/i3D2
SMIL [2H]C([2H])(O)[C@H]1COC(C)(C)O1
IUPAC navn dideuterio-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Molekylvægt (g/mol) 134.17
Formel vægt 134.0912