Få mere at vide
(S)-1-tert-Butylamino-2,3-propanediol, TRC
Artikelnummer. 30332829
Skift visning
Mængde:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30332829 1 g
30332513 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30332829 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6921551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S)-3-[(1,1-Dimethylethyl)amino]propane-1,2-diol
CAS 30315-46-9
Molekylær formel C7 H17 N O2
Mængde 1 g
Synonym 1,2-Propanediol, 3-[(1,1-dimethylethyl)amino]-, (2S)-, 1,2-Propanediol, 3-(tert-butylamino)-, (S)-(-)- (8CI), 1,2-Propanediol, 3-[(1,1-dimethylethyl)amino]-, (S)-, (S)-1-tert-Butylamino-2,3-propanediol, (S)-3-(tert-Butylamino)-1,2-propanediol
InChI formel InChI=1S/C7H17NO2/c1-7(2,3)8-4-6(10)5-9/h6,8-10H,4-5H2,1-3H3/t6-/m0/s1
SMIL CC(C)(C)NC[C@H](O)CO
IUPAC navn (2S)-3-(tert-butylamino)propane-1,2-diol
Molekylvægt (g/mol) 147.215
Formel vægt 147.1259