missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-(-)-1-Phenylpropylamine, 98%, BLD Pharm™

Artikelnummer. 30979872
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30979872 10 g
30974765 25 g
30976652 5 g
30969879 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30979872 Supplier BLD Pharm Supplier No. BD1136610g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale (S)-(-)-1-Phenylpropylamine
CAS 3789-59-1
Molekylær formel C9H13N
MDL nummer MFCD00082356
Mængde 10 g
SMIL CC[C@@H](C1=CC=CC=C1)N
Molekylvægt (g/mol) 135.21
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.