Learn More
Alfa Aesar™ (S)-(+)-1-Fmoc-3-pyrrolidinol, 95%
Brand: Alfa Aesar™ H57321.MD
Additional Details : CAS Number : 215178-38-4 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
215178-38-4 | |
309.365 | |
KPDNZJQVYQCDJW-ZDUSSCGKSA-N | |
16217955 | |
C1CN(CC1O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C19H19NO3 | |
MFCD01317842 | |
s-+-1-fmoc-3-hydroxypyrrolidine, s-+-1-fmoc-3-pyrrolidinol, s-1-fmoc-3-pyrrolidinol, 9h-fluoren-9-ylmethyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-1-n-fmoc-3-hydroxy-pyrrolidine, pubchem13966, ksc547a2h, s-1-9-fluorenylmethoxycarbonyl-3-pyrrolidinol, 9h-fluoren-9-ylmethyl 3s-3-hydroxy-1-pyrrolidinecarboxylate | |
9H-fluoren-9-ylmethyl (3S)-3-hydroxypyrrolidine-1-carboxylate |
Specifications
(S)-(+)-1-Fmoc-3-pyrrolidinol | |
250mg | |
C19H19NO3 | |
s-+-1-fmoc-3-hydroxypyrrolidine, s-+-1-fmoc-3-pyrrolidinol, s-1-fmoc-3-pyrrolidinol, 9h-fluoren-9-ylmethyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-1-n-fmoc-3-hydroxy-pyrrolidine, pubchem13966, ksc547a2h, s-1-9-fluorenylmethoxycarbonyl-3-pyrrolidinol, 9h-fluoren-9-ylmethyl 3s-3-hydroxy-1-pyrrolidinecarboxylate | |
C1CN(CC1O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
309.365 | |
309.36 |
106°C to 110°C | |
215178-38-4 | |
MFCD01317842 | |
KPDNZJQVYQCDJW-ZDUSSCGKSA-N | |
9H-fluoren-9-ylmethyl (3S)-3-hydroxypyrrolidine-1-carboxylate | |
16217955 | |
95% |