Learn More
(S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 225920-05-8 |
| Molecular Formula | C10H8F6O |
| MDL Number | MFCD03093011 |
| Quantity | 25 g |
| SMILES | C[C@H](O)C1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1 |
| Molecular Weight (g/mol) | 258.16 |
| Percent Purity | 96% |
| Chemical Name or Material | (S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol |