missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-[2-(Diphenylphosphino)phenyl]ethylamine, 97+%, Thermo Scientific Chemicals

Catalog Number p-7040362
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
913196-43-7
C20H20NP
305.36
MFCD17013987
KPFJIPHGQGHIMM-XISACWJONA-N
s-1-2-diphenylphosphino phenyl ethanamine, s-1-2-diphenylphosphino phenyl ethylamine, s---1-2-diphenylphosphino phenyl ethylamine, 1s-1-2-diphenylphosphanyl phenyl ethanamine, 2-1-aminoethyl triphenylphosphine, s-alpha-methyl-2-diphenylphosphino benzylamine, s-1-2-diphenylphosphanyl phenyl ethan-1-amine, 1s-1-2-diphenylphosphanyl phenyl ethan-1-amine
57514668
(1S)-1-(2-diphenylphosphanylphenyl)ethanamine
C[C@H](N)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
913196-43-7
C20H20NP
305.36
MFCD17013987
KPFJIPHGQGHIMM-XISACWJONA-N
s-1-2-diphenylphosphino phenyl ethanamine, s-1-2-diphenylphosphino phenyl ethylamine, s---1-2-diphenylphosphino phenyl ethylamine, 1s-1-2-diphenylphosphanyl phenyl ethanamine, 2-1-aminoethyl triphenylphosphine, s-alpha-methyl-2-diphenylphosphino benzylamine, s-1-2-diphenylphosphanyl phenyl ethan-1-amine, 1s-1-2-diphenylphosphanyl phenyl ethan-1-amine
57514668
(1S)-1-(2-diphenylphosphanylphenyl)ethanamine
C[C@H](N)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 81°C
Quantity 250 mg
Sensitivity Air Sensitive
Optical Rotation −55° (c=0.5 in Chloroform)
Formula Weight 305.35
Percent Purity ≥97%
Chemical Name or Material (S)-(-)-1-[2-(Diphenylphosphino)phenyl]ethylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'