Få mere at vide
(S)-1-(2-Chloroacetyl)pyrrolidine-2-carboxamide, TRC
Artikelnummer. 30137709
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137709 1 g
30134974 500 mg
30008596 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137709 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3638651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 214398-99-9
Molekylær formel C7 H11 Cl N2 O2
Molekylvægt (g/mol) 190.63
Synonym 2-Pyrrolidinecarboxamide, 1-(2-chloroacetyl)-, (2S)-, 2-Pyrrolidinecarboxamide, 1-(chloroacetyl)-, (2S)- (9CI), (2S)-1-(2-Chloroacetyl)-2-pyrrolidinecarboxamide, (S)-1-(2-Chloroacetyl)pyrrolidine-2-carboxamide, 1-(2-Chloroacetyl)-2-(S)-pyrrolidine carboxamide, 1-(Chloroacetyl)-L-prolinamide
IUPAC navn (2S)-1-(2-chloroacetyl)pyrrolidine-2-carboxamide
SMIL NC(=O)[C@@H]1CCCN1C(=O)CCl
Tekniske data
Kemisk navn eller materiale (S)-1-(2-Chloroacetyl)pyrrolidine-2-carboxamide
Renhedsgrad noter HPLC
InChI formel InChI=1S/C7H11ClN2O2/c8-4-6(11)10-3-1-2-5(10)7(9)12/h5H,1-4H2,(H2,9,12)/t5-/m0/s1
Mængde 1 g
Formel vægt 190.0509
Procent renhed >95%