Få mere at vide
Tocris Bioscience™ RWJ 67657
Beskrivelse
Selective p38α and p38β inhibitor (IC50 values are 1 and 11 μM respectively) that displays no activity at p38γ, p38δ and a range of other kinases. Potently inhibits TNF-α and IL-1β release (IC50 values are 3 and 11 nM respectively) and suppresses HIV-1 replication in T cells in vitro. Displays cardioprotective and anti-inflammatory activity in vivo. Orally active.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | RWJ 67657 |
| CAS | 215303-72-3 |
| Mængde | 10 mg |
| Molekylær formel | C27H24FN3O |
| Formel vægt | 425.5 |
| Renhed | >98% |
| Molekylvægt (g/mol) | 425.507 |