Få mere at vide
Ruxolitinib-amide, TRC
Artikelnummer. 30414716
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414716 10 mg
30410939 1 mg
30408999 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30414716 Leverandør Toronto Research Chemicals Leverandørnr. TRCR70198510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1911644-32-0
Molekylær formel C17H20N6O
Molekylvægt (g/mol) 324.38
Synonym 3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanamide
IUPAC navn 3-cyclopentyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]propanamide
SMIL NC(=O)CC(C1CCCC1)n2cc(cn2)c3ncnc4[nH]ccc34
Tekniske data
Kemisk navn eller materiale Ruxolitinib-amide
Mængde 10 mg
InChI formel InChI=1S/C17H20N6O/c18-15(24)7-14(11-3-1-2-4-11)23-9-12(8-22-23)16-13-5-6-19-17(13)21-10-20-16/h5-6,8-11,14H,1-4,7H2,(H2,18,24)(H,19,20,21)
Formel vægt 324.1699