missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ruthenium, 5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet

Brand:  Alfa Aesar™ 045439.14

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15483985

  • 1133.61 DKK / 25g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]

Specifications

Ruthenium
7440-18-8
MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Odorless
25g
5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet
Ru
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
Black
Videos
SDS Documents