missing translation for 'onlineSavingsMsg'
Få mere at vide

Rupintrivir, TRC

Artikelnummer. 30409389
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.25 mg
2.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409389 2.5 mg
30412827 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30409389 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7017002.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 223537-30-2
Molekylær formel C31H39FN4O7
Molekylvægt (g/mol) 598.66
Synonym (2E,4S)-4-[[(2R,5S)-2-[(4-Fluorophenyl)methyl]-6-methyl-5-[[(5-methyl-3-isoxazolyl)carbonyl]amino]-1,4-dioxoheptyl]amino]-5-[(3S)-2-oxo-3-pyrrolidinyl]-2-pentenoic Acid, AG 7088
IUPAC navn ethyl (E,4S)-4-[[(2R,5S)-2-[(4-fluorophenyl)methyl]-6-methyl-5-[(5-methyl-1,2-oxazole-3-carbonyl)amino]-4-oxoheptanoyl]amino]-5-[(3S)-2-oxopyrrolidin-3-yl]pent-2-enoate
SMIL CCOC(=O)\C=C\[C@H](C[C@@H]1CCNC1=O)NC(=O)[C@@H](CC(=O)[C@@H](NC(=O)c2cc(C)on2)C(C)C)Cc3ccc(F)cc3

Tekniske data

Kemisk navn eller materiale Rupintrivir
Mængde 2.5 mg
InChI formel InChI=1S/C31H39FN4O7/c1-5-42-27(38)11-10-24(16-21-12-13-33-29(21)39)34-30(40)22(15-20-6-8-23(32)9-7-20)17-26(37)28(18(2)3)35-31(41)25-14-19(4)43-36-25/h6-11,14,18,21-22,24,28H,5,12-13,15-17H2,1-4H3,(H,33,39)(H,34,40)(H,35,41)/b11-10+/t21-,22+,24+,28-/m0/s1
Formel vægt 598.2803
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.