Få mere at vide
Rupatadine Fumarate, TRC
Artikelnummer. 30006122
Skift visning
Mængde:
5 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006122 5 mg
30010915 100 mg
30012147 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006122 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7016505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 182349-12-8
Molekylær formel C26 H26 Cl N3 . C4 H4 O4
Molekylvægt (g/mol) 532.03
Synonym 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 8-chloro-6,11-dihydro-11-[1-[(5-methyl-3-pyridinyl)methyl]-4-piperidinylidene]-, (2E)-2-butenedioate (1:1), 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 8-chloro-6,11-dihydro-11-[1-[(5-methyl-3-pyridinyl)methyl]-4-piperidinylidene]-, (E)-2-butenedioate (1:1), Rupatadine fumarate
IUPAC navn (E)-but-2-enedioic acid;8-chloro-11-[1-[(5-methylpyridin-3-yl)methyl]piperidin-4-ylidene]-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMIL Cc1cncc(CN2CCC(=C3c4ccc(Cl)cc4CCc5cccnc35)CC2)c1.OC(=O)\C=C\C(=O)O
Tekniske data
Kemisk navn eller materiale Rupatadine Fumarate
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C26H26ClN3.C4H4O4/c1-18-13-19(16-28-15-18)17-30-11-8-20(9-12-30)25-24-7-6-23(27)14-22(24)5-4-21-3-2-10-29-26(21)25;5-3(6)1-2-4(7)8/h2-3,6-7,10,13-16H,4-5,8-9,11-12,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+
Formel vægt 531.1925
Procent renhed >95%