Få mere at vide
Rupatadine-d4 Fumarate, TRC
Artikelnummer. 30410443
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30410443 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410443 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7016522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rupatadine-d4 Fumarate
CAS 1795153-63-7
Molekylær formel C30H26D4ClN3O4
Mængde 2.5 mg
Synonym 8-Chloro-6,11-dihydro-11-[1-[(5-methyl-3-pyridinyl)methyl]-4-piperidinylidene-d4]- 5H-benzo[5,6]cyclohepta[1,2-b]pyridine (2E)-2-Butenedioate, Rupafin-d4 Fumarate
InChI formel InChI=1S/C26H26ClN3.C4H4O4/c1-18-13-19(16-28-15-18)17-30-11-8-20(9-12-30)25-24-7-6-23(27)14-22(24)5-4-21-3-2-10-29-26(21)25;5-3(6)1-2-4(7)8/h2-3,6-7,10,13-16H,4-5,8-9,11-12,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/i11D2,12D2;
SMIL [2H]C1(CC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC(N1CC5=CN=CC(=C5)C)([2H])[2H])[2H].C(=C/C(=O)O)\C(=O)O
IUPAC navn (E)-but-2-enedioic acid;8-chloro-11-[2,2,6,6-tetradeuterio-1-[(5-methylpyridin-3-yl)methyl]piperidin-4-ylidene]-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molekylvægt (g/mol) 536.05
Formel vægt 535.2176