Få mere at vide
Rufinamide USP Related Compound B, Mikromol™
Artikelnummer. 30461213
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461213 4 x 25 mg
30049229 25 mg
30052579 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461213 Leverandør Mikromol Leverandørnr. MM3338.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Rufinamide USP Related Compound B, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl 1-(2,6-difluorobenzyl)-1H-1,2,3-triazole-4-carboxylate
CAS 217448-86-7
Analyt- eller komponentnavne Methyl 1-(2,6-difluorobenzyl)-1H-1,2,3-triazole-4-carboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C11 H9 F2 N3 O2
Certificeringer/compliance ISO 17025
Synonym Methyl 1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carboxylate, Rufinamide Related Compound B
InChI formel InChI=1S/C11H9F2N3O2/c1-18-11(17)10-6-16(15-14-10)5-7-8(12)3-2-4-9(7)13/h2-4,6H,5H2,1H3
Løsningstype Neat
SMIL COC(=O)c1cn(Cc2c(F)cccc2F)nn1
IUPAC navn methyl 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxylate
Molekylvægt (g/mol) 253.2
Mængde 4 x 25 mg
Formel vægt 253.0663 g/mol