Få mere at vide
Rufinamide-5-Carboxamide, TRC
Artikelnummer. 30410978
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30410978 5 mg -
30425535 2.5 mg 2.5mg
30418929 0.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30410978 Leverandør Toronto Research Chemicals Leverandørnr. TRCR7014705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1622904-99-7
Molekylær formel C10H8F2N4O
Molekylvægt (g/mol) 238.19
Synonym Rufinamide Impurity 3
IUPAC navn 3-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
SMIL NC(=O)c1cnnn1Cc2c(F)cccc2F
Tekniske data
Kemisk navn eller materiale Rufinamide-5-Carboxamide
Mængde 5 mg
InChI formel InChI=1S/C10H8F2N4O/c11-7-2-1-3-8(12)6(7)5-16-9(10(13)17)4-14-15-16/h1-4H,5H2,(H2,13,17)
Formel vægt 238.0666