Learn More
MedChemExpress Rubitecan
Product Code. 16451987
Encompass_Preferred
Change view
Click to view available options
50 mg
Product Code.
16451987 50 mg
1 options
Product Code. 16451987 Supplier MedChemExpress Supplier No. HY13744
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Rubitecan (RFS 2000), a Camptothecin derivative, is an orally active topoisomerase I inhibitor with broad antitumor activity, and induces protein-linked DNA single-strand breaks, thereby blocking DNA and RNA synthesis in dividing cells.
Specifications
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 393.35 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 91421-42-0
Product Type Natural Products
Purity 98.07%
Solubility DMSO : 125 mg/mL (317.78 mM∣Need ultrasonic)
Synonym RFS 2000∣ 9-Nitrocamptothecin
Target Topoisomerase
Quantity 50 mg