missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Rubicon Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30461C
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Rubicon Polyclonal antibody specifically detects Rubicon in Human samples. It is validated for Western Blot
Tekniske data
Rubicon | |
Polyclonal | |
Western Blot | |
Baron, Beclin 1-interacting protein, Beclin-1 Associated RUN Domain Containing Protein, KIAA0226, Rubicon, RUN And Cysteine Rich Domain Containing Beclin 1 Interacting Protein, RUN Domain And Cysteine-Rich Domain Containing, Beclin 1-Interacting Protein, Run Domain Beclin-1-Interacting And Cysteine-Rich Domain-Containing Protein, rundataxin, SCAR15 | |
The immunogen for this product maps to a region between residue 922 and 972 of human Rubicon using the numbering given in entry NP_055502.1 (GeneID 9711). | |
0.1 mL | |
Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
9711 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Rubicon Antibody, DyLight 650, Novus Biologicals™ > 0.1 mL; DyLight 650
Ser du en mulighed for forbedring?Del en indholdskorrektion