Learn More
Fisherbrand™ Rubber Tubing
For transferring liquids, gases and dilute acids.
164.00 DKK - 1840.00 DKK
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | |||
11895512
|
Fisherbrand™
|
14 mm | 10 mm | 2 mm |
324.00 DKK
10m |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
11507572
|
Fisherbrand™
|
24 mm | 10 mm | 7 mm |
1115.00 DKK
10m |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
11805522
|
Fisherbrand™
|
17 mm | 12.5 mm | 2.5 mm |
352.00 DKK
10m |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
11815522
|
Fisherbrand™
|
22.5 mm | 16 mm | 3.25 mm |
630.00 DKK
10m |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
11825522
|
Fisherbrand™
|
26.5 mm | 20 mm | 3.25 mm |
725.00 DKK
10m |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
11835522
|
Fisherbrand™
|
31.5 mm | 25 mm | 3.25 mm |
1185.00 DKK
10m |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
11865512
|
Fisherbrand™
|
8 mm | 5 mm | 1.5 mm |
164.00 DKK
10m |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
11855512
|
Fisherbrand™
|
4.5 mm | 3 mm | 0.75 mm |
172.00 DKK
10m |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
11587562
|
Fisherbrand™
|
16.5 mm | 6.5 mm | 5 mm |
525.00 DKK
10m |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
11885512
|
Fisherbrand™
|
12 mm | 8 mm | 2 mm |
227.00 DKK
10m |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
11875512
|
Fisherbrand™
|
9.5 mm | 6.5 mm | 1.5 mm |
178.00 DKK
10m |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
11517572
|
Fisherbrand™
|
29.5 mm | 12.5 mm | 8.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15199654
|
Fisherbrand™
|
9.5 mm | 6.5 mm | 1.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Higher resilience than most other polymers
- Temperature range: -40°C to 70°C
- Light and flexible
- Not suitable for use with petroleum oils and hydrocarbon solvents (see neoprene tubing)
Specifications
Rubber | |
10 m |