Learn More
Saint-Gobain Rubber Small Volume Pacifier
Use this small volume pacifier for a variety of applications. Saint Gobain Performance Plastics™ Rubber Small Volume Pacifier are compatible with a wide range of pipettes.
139.00 DKK - 431.00 DKK
Product Code | Brand | Color | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Capacity (Metric) | Price | Quantity & Availability | ||||
11704508
|
Saint-Gobain
10101310 |
Blue | 10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11784498
|
Saint-Gobain
10101110 |
White | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11774498
|
Saint-Gobain
10101010 |
Blue | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- For liquid handling
Specifications
10/Pk. | |
Small Volume Pacifier | |
Rubber |