Learn More
Deutsch Neumann™ Rubber Policemen
Provides for quick and easy transfer of residues to glass surfaces. Deutsch Neumann™ Rubber Policemen are manufactured from natural rubber.
Brand:  Deutsch Neumann™ 2250001
Additional Details : Weight : 0.00090kg
 View more versions of this product
Product Code. 10028230
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • 5mm hole for insert
  • Available in grey or red color
  • Durometer Shore A 40
  • Excellent tensile strength and resilience
Specifications
Specifications
Rubber
Gray