Tekniske data
| Mængde | 0.1 mL |
| Indhold og opbevaring | Store at -80°C. Avoid freeze/thaw cycles. |
| Mål | RTKN2 |
| Synonym | bA531F24.1, DKFZp686J10120, Em:AC024597.2, FLJ39352, PH domain-containing family K member 1, pleckstrin homology domain containing, family K member 1, Pleckstrin homology domain-containing family K member 1, PLEKHK1, rhotekin 2, rhotekin-2 |
| Gen symbol | RTKN2 |