Få mere at vide
(RS)-N-(4-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride, Mikromol™
Beskrivelse
(RS)-N-(4-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (RS)-N-(4-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride |
| CAS | 35891-99-7 |
| Analyt- eller komponentnavne | (RS)-N-(4-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H20 N2 O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Prilocaine Hydrochloride Imp. E (EP), Prilocaine Imp. E (EP), Prilocaine USP Related Compound B, Prilocaine USP RC B, (RS)-N-(4-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride, Prilocaine Hydrochloride Imp. E (EP) as Hydrochloride, Prilocaine Related Compound B as Hydrochloride, Prilocaine Hydrochloride Impurity E as Hydrochloride, Prilocaine Impurity E as Hydrochloride |
| InChI formel | InChI=1S/C13H20N2O.ClH/c1-4-9-14-11(3)13(16)15-12-7-5-10(2)6-8-12;/h5-8,11,14H,4,9H2,1-3H3,(H,15,16);1H |
| SMIL | Cl.CCCNC(C)C(=O)Nc1ccc(C)cc1 |
| IUPAC navn | N-(4-methylphenyl)-2-(propylamino)propanamide;hydrochloride |
| Vis mere |